missing translation for 'onlineSavingsMsg'
Learn More

Acetyl Chloride, Technical, Fisher Chemical

Product Code. 10672241 Shop All Fisher Chemical Products
Change view
Click to view available options
5AMP, Ampoule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10672241 5AMP, Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10672241 Supplier Fisher Chemical Supplier No. A/0881/99
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-36-5
Melting Point -112°C
Color Colorless
Boiling Point 51°C
Packaging Ampoule
Molecular Formula C2H3ClO
MDL Number 719
Quantity 5 Ampules
Formula Weight 78.5g/mol
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.