missing translation for 'onlineSavingsMsg'
Learn More

Acetyl bromide, 98%

Product Code. 10205970 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10205970 500 mL 500mL
10734651 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 10205970 Supplier Thermo Scientific Acros Supplier No. 159405000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 506-96-7 | C2H3BrO | 122.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 506-96-7
Molecular Formula C2H3BrO
Molecular Weight (g/mol) 122.95
MDL Number MFCD00000114
InChI Key FXXACINHVKSMDR-UHFFFAOYSA-N
Synonym ethanoyl bromide, acetylbromide, acetic acid, bromide, ch3cobr, unii-o18v5xyo0g, hsdb 663, o18v5xyo0g, bromideacetyl, acbr, acetyl bromide
PubChem CID 10482
IUPAC Name acetyl bromide
SMILES CC(=O)Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -96°C
Color Colorless to Yellow
Density 1.6600g/mL
Boiling Point 75°C to 77°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3COBr
Refractive Index 1.4485 to 1.4505
Quantity 500 mL
Beilstein 02, 174
Fieser 06,9; 13,208
Merck Index 15, 77
Specific Gravity 1.66
Solubility Information Solubility in water: reacts. Other solubilities: reacts with alcohol and acetone,soluble in ether,chloroform and benzene
Formula Weight 122.95
Percent Purity 98%
Physical Form Fuming Liquid
Chemical Name or Material Acetyl bromide, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.