missing translation for 'onlineSavingsMsg'
Learn More

Acetrizoic acid, MedChemExpress

Product Code. 16408756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16408756 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16408756 Supplier Medchem Express Supplier No. HYB1406
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Acetrizoic acid is a molecule used as a contrast medium.

Acetrizoic acid is a molecule used as a contrast medium.

Specifications

Chemical Name or Material Acetrizoic acid
CAS 85-36-9
Color White
Purity Grade Notes Research
Molecular Formula C9H6I3NO3
Quantity 100 mg
Solubility Information DMSO : ≥ 5.6 mg/mL (10.06 mM)
SMILES O=C(O)C1=C(I)C=C(I)C(NC(C)=O)=C1I
Molecular Weight (g/mol) 556.86
Formula Weight 556.86
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.