missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetyl chloride, 97%

Product Code. 10407412 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10407412

Brand: Thermo Scientific Acros 370070250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13831-31-7 | C4H5ClO3 | 136.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13831-31-7
Molecular Formula C4H5ClO3
Molecular Weight (g/mol) 136.53
MDL Number MFCD00011535
InChI Key HZDNNJABYXNPPV-UHFFFAOYSA-N
Synonym acetoxyacetyl chloride, 2-acetoxyacetyl chloride, unii-z4s19y2f8s, acetoxyacetylchloride, acetyl chloride, acetyloxy, acetoxyacetic acid chloride, chlorocarbonyl methyl acetate, acetoxy acetylchloride, acetyloxyacetyl chloride, pubchem20920
PubChem CID 26297
IUPAC Name (2-chloro-2-oxoethyl) acetate
SMILES CC(=O)OCC(=O)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 64°C to 68°C (25.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2CH2COCl
Refractive Index 1.4255 to 1.4275
Quantity 25 g
Specific Gravity 1.27
Formula Weight 136.53
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Acetoxyacetyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Acetoxyacetyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.