missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetone, 97%

Product Code. 15414077
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15414077 10 mL 10mL
15424077 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15414077 Supplier Thermo Scientific Alfa Aesar Supplier No. H31346.AB
  • ONLINE EXCLUSIVE PRICE
    £53.50
    £48.65 / 10mL
    Save £4.85
    9% Off
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 592-20-1 | C5H8O3 | 116.116 g/mol

Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetoxyacetone is an building block in the synthesis of various pharmaceutical and organic compounds.

Solubility
Soluble in diethyl ether, ethyl acetate and dichloromethane.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-20-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00042848
InChI Key DBERHVIZRVGDFO-UHFFFAOYSA-N
Synonym acetoxyacetone, acetonyl acetate, acetol acetate, 2-propanone, 1-acetyloxy, acetoxypropanone, o-acetylacetol, 1-acetoxyacetone, acetylmethyl acetate, acetoxy-2-propanone, 1-acetoxy-2-propanone
PubChem CID 11593
IUPAC Name 2-oxopropyl acetate
SMILES CC(=O)COC(=O)C

Specifications

Density 1.075
Boiling Point 174°C to 176°C
Flash Point 71°C (160°F)
Refractive Index 1.417
Quantity 10 mL
Solubility Information Soluble in diethyl ether,ethyl acetate and dichloromethane.
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material Acetoxyacetone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.