missing translation for 'onlineSavingsMsg'
Learn More

Acetoxyacetone 95%, Thermo Scientific™

Product Code. 10559983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10559983 50 mL Glass bottle 50mL
10549983 10 mL Glass bottle 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10559983 Supplier Thermo Scientific Acros Supplier No. 389680500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-20-1
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00042848
InChI Key DBERHVIZRVGDFO-UHFFFAOYSA-N
Synonym acetoxyacetone, acetonyl acetate, acetol acetate, 2-propanone, 1-acetyloxy, acetoxypropanone, o-acetylacetol, 1-acetoxyacetone, acetylmethyl acetate, acetoxy-2-propanone, 1-acetoxy-2-propanone
PubChem CID 11593
IUPAC Name 2-oxopropyl acetate
SMILES CC(=O)COC(=O)C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Amber to Yellow
Density 1.0750g/mL
Boiling Point 179.0°C
Flash Point 75°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula CH3C(O)OCH2C(O)CH3
Refractive Index 1.4140 to 1.4160
Quantity 50 mL
Specific Gravity 1.075
Solubility Information Solubility in water: slightly soluble.
Formula Weight 116.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Acetoxyacetone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.