missing translation for 'onlineSavingsMsg'
Learn More

Acetovanillone, 98%

Product Code. 10152940 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10152940 100 g Glass bottle 100g
10070540 25 g Glass bottle 25g
10723465 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10152940 Supplier Thermo Scientific Acros Supplier No. 102421000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 498-02-2 | C9H10O3 | 166.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 498-02-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00008747
InChI Key DFYRUELUNQRZTB-UHFFFAOYSA-N
Synonym acetovanillone, apocynin, 4'-hydroxy-3'-methoxyacetophenone, 1-4-hydroxy-3-methoxyphenyl ethanone, acetoguaiacone, acetoguaiacon, apocynine, 4-acetyl-2-methoxyphenol, acetovanilone, acetovanyllon
PubChem CID 2214
ChEBI CHEBI:2781
IUPAC Name 1-(4-hydroxy-3-methoxyphenyl)ethanone
SMILES COC1=CC(=CC=C1O)C(C)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 112.0°C to 116.0°C
Color Yellow to Brown
Boiling Point 295.0°C to 300.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H3(OCH3)COCH3
Quantity 100 g
Beilstein 08, 272
Merck Index 15, 728
Solubility Information Solubility in water: soluble in hot water. Other solubilities: soluble in ethanol,acetone,benzene,very soluble in diethyl ether,chloroform,practically insoluble in petroleum ether
Formula Weight 166.18
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Acetovanillone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.