missing translation for 'onlineSavingsMsg'
Learn More

Acetophenone, TRC

Product Code. 30136824
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136824 25 g 25g
17984235 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136824 Supplier Toronto Research Chemicals Supplier No. TRCA16401525G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-86-2
Molecular Formula C8 H8 O
Molecular Weight (g/mol) 120.15
Synonym Acetophenone (8CI), 1-Phenylethanone, 1-Feniletanone, 1-Phenyl-1-ethanone, Acetophenon, Acetylbenzene, Benzoylmethane, Hypnon, Hypnone, Methyl phenyl ketone, NSC 7635, NSC 98542, Phenyl methyl ketone
IUPAC Name 1-phenylethanone
SMILES CC(=O)c1ccccc1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3
Formula Weight 120.0575
Percent Purity >95%
Chemical Name or Material Acetophenone
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.