missing translation for 'onlineSavingsMsg'
Learn More

Acetophenazine Dimaleate, TRC

Product Code. 30134090
Change view
Click to view available options
Quantity:
100 mg
200 mg
1 g
Unit Size:
100mg
1g
200mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134090 200 mg 200mg
30135541 1 g 1g
30005786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134090 Supplier Toronto Research Chemicals Supplier No. TRCA163930200MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5714-00-1
Molecular Formula C23 H29 N3 O2 S . 2 C4 H4 O4
Molecular Weight (g/mol) 643.7
Synonym Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2)Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (2Z)-2-butenedioate (1:2) (salt) (9CI), Ethanone, 1-[10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]-, (Z)-2-butenedioate (1:2) (salt), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, dimaleate (6CI), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, maleate (1:2) (salt) (8CI), Ketone, 10-[3-[4-(2-hydroxyethyl)-1-piperazinyl]propyl]phenothiazin-2-yl methyl, maleate (7CI), Acetophenazine dimaleate, Acetophenazine maleate, NSC 169180, NSC 70600, Phentoxate, Sch 6673, Tindal, Tindala
IUPAC Name (Z)-but-2-enedioic acid;1-[10-[3-[4-(2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]ethanone
SMILES CC(=O)c1ccc2Sc3ccccc3N(CCCN4CCN(CCO)CC4)c2c1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O

Specifications

Quantity 200 mg
InChI Formula InChI=1S/C23H29N3O2S.2C4H4O4/c1-18(28)19-7-8-23-21(17-19)26(20-5-2-3-6-22(20)29-23)10-4-9-24-11-13-25(14-12-24)15-16-27;2*5-3(6)1-2-4(7)8/h2-3,5-8,17,27H,4,9-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Formula Weight 643.22
Chemical Name or Material Acetophenazine Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.