missing translation for 'onlineSavingsMsg'
Learn More

Acetonylacetone, 97%, ACROS Organics™

Brand:  Acros Organics 102400050

Code : 29

Additional Details : CAS Number : 110-13-4 Weight : 0.05487kg


Packaging Glass bottle
Quantity 5mL

 View more versions of this product

Product Code. 10050720

  • £14.35 / 5mL

Estimated Shipment: 16-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

110-13-4
114.14
OJVAMHKKJGICOG-UHFFFAOYSA-N
8035
hexane-2,5-dione
C6H10O2
MFCD00008792
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
CHEBI:85014
CC(=O)CCC(=O)C

Specifications

Acetonylacetone
78°C
Glass bottle
0.973
185.0°C to 193.0°C
-6.0°C
97%
96.0
97%
CH3COCH2CH2COCH3
01,788
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
OJVAMHKKJGICOG-UHFFFAOYSA-N
hexane-2,5-dione
8035
114.14
97%
0.9730g/mL
Authentic
1.4250 to 1.4270
1.43 mPa.s (25°C)
Yellow to Brown
5mL
110-13-4
100.0
C6H10O2
MFCD00008792
15,7
Solubility in water: miscible. Other solubilities: miscible with alcohol, ether, ethanol and, diethyl ether, miscible in organic solvents
CC(=O)CCC(=O)C
114.14
CHEBI:85014
Liquid
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetonylacetone

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates