missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile solution, Honeywell™ Riedel-de-Haën™

Product Code. 15645850
Change view
Click to view available options
Quantity:
20 L
Packaging:
Metal Can
Unit Size:
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15645850 20 L Metal Can 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15645850 Supplier Honeywell-Riedel-de Haen Supplier No. 56542320L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 75-05-8
Density 0.782g/mL at 25°C
Quantity 20 L
Flash Point 6°C (42.8°F)
Packaging Metal Can
Molecular Formula C2H3N
Linear Formula CH3CN
MDL Number MFCD00001878
UN Number UN1648
Beilstein 4124158
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05g/mol
Chemical Name or Material Acetonitrile solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.