missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC), Honeywell Riedel-de Haën™

Product Code. 15664670
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15664670 1 L Glass Bottle 1 L
15674670 2.5 L Glass Bottle 2.5 L
15694670 6 x 1 L Glass Bottle Pack of 6
15684670 4 x 2.5 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15664670 Supplier Honeywell-Chromasolv Supplier No. 348811L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R CHROMASOLV™, for liquid chromatography,≥99.8% (GC)

  • For GC headspace applications
  • Meets USP, Ph. Eur. and ICH requirements
  • GC headspace application tested
  • Packed under inert gas for longer shelf life
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C (literature)
CAS Max % 100.0000%
Density 0.786 g/mL at 25 °C(lit.)
Quantity 1 L
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CH3CN
UN Number UN1648
Beilstein 741857
Boiling Point 81.6°C
Formula Weight 41.05
Percent Purity >99.8%
Grade Chromatography
Chemical Name or Material Acetonitrile, R CHROMASOLV™, for liquid chromatography, ≥99.8% (GC)
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302 + H312 + H332-Harmful if swallowed, in contact with skin or if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.