missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™

Product Code. 15623880
Change view
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Unit Size:
155kg
Pack of 4
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15623880 6 x 1 L Glass Bottle Pack of 6
15613880 4 x 2.5 L Glass Bottle Pack of 4
15673870 155 kg Metal Drum 155kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15623880 Supplier Honeywell-Riedel-de Haen Supplier No. 330196X1L
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss. p.a., ACS Reagent, Reag. Ph. Eur.,≥99.5% (GC)

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Acidity Free acid (as CH3COOH): ≤0.001%
Alkalinity Free alkali (as NH3): ≤0.001%
Density 0.786g/mL at 25°C
Quantity 6 x 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Packaging Glass Bottle
Refractive Index n20/D 1.3435 to 1.3445
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
UN Number UN1648
Beilstein 741857
Boiling Point 82°C
Formula Weight 41.05g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Chemical Name or Material Acetonitrile, Reag. Ph. Eur.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.