missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, puriss. p.a. ACS Reagent, Honeywell

Catalog Number 15683870
Click to view available options
Quantity:
1 L
2.5 L
25 L
This item is not returnable. View return policy
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
This item is not returnable. View return policy
75-05-8
C2H3N
41.053
MFCD00001878
WEVYAHXRMPXWCK-UHFFFAOYSA-N
methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
6342
CHEBI:38472
acetonitrile
CC#N
Linear Formula CH3CN
UN Number 1648
Beilstein 741857
Formula Weight 41.05g/mol
Percent Purity ≥99.5% (GC)
Grade ACS/puriss. p.a.
Density 0.786g/mL at 25°C
Quantity 1 L
Explosion Limit 0.16
Flash Point 2°C (35.6°F)
Ignition Point 973°F
Vapor Pressure 72.8mmHg (20°C)
Chemical Name or Material Acetonitrile, Reag. Ph. Eur.
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'