missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Low-Water, BAKER BIO-ANALYZED, J.T. Baker™

Product Code. 15528504 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15528504 4 L Glass 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15528504 Supplier JT Baker Supplier No. 901803

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carefully controlled low water levels and unsurpassed lot-to-lot consistency deliver better synthesis yields and coupling efficiencies.

BAKER BIO-ANALYZED products are a group of high purity solvents and reagents specially purified and carefully tested to assure suitability and reliability in nucleic acid or peptide synthesis.

Specifications

Melting Point -46°C
Absorbance Max. 0.05 at 200nm, Max. 0.01 at 220nm, Max. 0.010 at 254-400nm
CAS 75-05-8
Color Colorless
Appearance Passes test
Density 0.7822g/cm3 at 20°C
Quantity 4 L
Explosion Limit 4.4 - 16 Vol %
Flash Point 5°C
Ignition Point 525°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 1.0 ppm
Titratable Acid Max. 0.8μeq/g
Molecular Formula C2H3N
Titratable Base Max. 0.6μeq/g
Linear Formula CH3CN
Viscosity 0.39 mPas at 20°C
Water Max. 10 ppm
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Boiling Point 81°C
Solubility Information Fully miscible in water
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05g/mol
Percent Purity Min. 99.8%
Grade For laboratory, research or manufacturing use
Assay By GC, corrected for water
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.