missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, LC-MS Ultra CHROMASOLV™, Solstice

CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS

Brand:  Honeywell-Chromasolv 14261-2L

299.01 GBP valid until 2026-10-31
BEST PRICE on Promo! Use promo code "27992" to get your promotional price.



 View more versions of this product

Product Code. 15611400

  • £380.00 / 2 L
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
In Stock
This item is not returnable. View return policy

Description

Description

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles
TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Acetonitrile

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302+H312+H332-Harmful if swallowed, in contact with skin or if inhaled
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS Documents

Documents

Certificates
Promotions

Promotions

Promo
Expires: 2026-10-31
Big Savings for Better Separation
Promo Code: 27992
Save up to 60% on chromatography consumables.

Applying Promo and Adding...

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.