missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile-d{3}, 99.8% (Isotopic)

Product Code. 11379937
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
Unit Size:
10g
2g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379937 50 g 50g
11369937 2 g 2g
11389937 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11379937 Supplier Thermo Scientific Alfa Aesar Supplier No. 042264.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 2206-26-0 | C2H3N | 44.07 g/mol

Acetonitrile d3 is used as a solvent for NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2206-26-0
Molecular Formula C2H3N
Molecular Weight (g/mol) 44.07
MDL Number MFCD00001881
InChI Key WEVYAHXRMPXWCK-FIBGUPNXSA-N
Synonym acetonitrile-d3, 2h3 acetonitrile, cd3cn, acetonitrile-2,2,2-d3, methyl-d3 cyanide, trideuteroacetonitrile, acetonitrile-d3, 99.8 atom % d, acetonitrile-d3, ≥99.8 atom % d, acetonitrile-d isotopic 5g
PubChem CID 123151
IUPAC Name 2,2,2-trideuterioacetonitrile
SMILES [2H]C([2H])([2H])C#N

Specifications

Physical Form Liquid
Color Colorless
Density 0.844 g/mL
Flash Point 5°C (42°F)
Odor Pungent
Sensitivity Hygroscopic
Quantity 50 g
Boiling Point 81°C
UN Number UN1648
Merck Index 14,70
Solubility Information Miscible with water,ethanol,diethyl ether,acetone,benzene,methanol,methyl acetate,ethyl acetate,acetamide solutions,chloroform,carbon tetrachloride,ethylene chloride and unsaturated hydrocarbons. Immiscible with saturated hydrocarbons.
Formula Weight 44.08
Percent Purity 99.8%
Chemical Name or Material Acetonitrile-d3, Isotopic
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.