missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, CHROMASOLV™, for HPLC, gradient grade, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15664520
Click to view available options
Quantity:
1 L
100 mL
2.5 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
100mL
2.5 L
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15664520

Brand: Honeywell-Chromasolv 348516X1L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CHROMASOLV™, gradient grade, for HPLC,≥99.9%

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -48°C
Absorbance HPLC-gradient/254nm ≤0.5mAU
Density 0.786g/mL at 25°C
Boiling Point 81°C to 82°C
Quantity 6 x 1 L
Refractive Index n20/D 1.344
Linear Formula CH3CN
Vapor Pressure 72.8mmHg (20°C)
Beilstein 741857
Formula Weight 41.05g/mol
Percent Purity ≥99.9%
Grade CHROMASOLV™ Gradient
Chemical Name or Material Acetonitrile, for HPLC
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : acetonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.