missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, ChromAR™ for HPLC, Macron Fine Chemicals™

Product Code. 15518724
Change view
Click to view available options
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15518724 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15518724 Supplier Macron Fine Chemicals Supplier No. 671125
  • ONLINE EXCLUSIVE PRICE
    £272.00
    £240.83 / 2.5 L
    Save £31.17
    11% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For High Performance Liquid Chromatography

Specifications

CAS 75-05-8
Quantity 2.5 L
Molecular Formula C2H3N
MDL Number MFCD00001878
UN Number 1648
Synonym Methyl cyanide, Ethanoic acid nitrile, Ethanenitrile, Cyanomethane
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.