missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, Biotech Grade, dried (max. 10 ppm H2O), Honeywell Burdick & Jackson™

Product Code. 16675893
Click to view available options
:
7 L
This item is not returnable. View return policy

Product Code. 16675893

Brand: Honeywell Burdick & Jackson 657397L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Melting Point -46°C
CAS 75-05-8
Color Colorless
CAS Max % 1
Density 0.7857 g/cm3 (20°C)
Boiling Point 82°C (1013 hPa)
Quantity 7 L
Flash Point 6°C
Molecular Formula C2H3N
Linear Formula CH3CN
Beilstein 741857
Synonym ACN; Methyl cyanide
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
Grade Anhydrous
Chemical Name or Material Acetonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.