missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, BAKER ANALYZED™ for LC-MS, J.T. Baker™

Product Code. 12759600 Shop All J.T.Baker Products
Change view
Click to view available options
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12759600 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12759600 Supplier JT Baker Supplier No. 9821.1000GL
  • ONLINE EXCLUSIVE PRICE
    £461.69
    £372.02 / 1 L
    Save £89.67
    19% Off
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

For liquid chromatography - mass spectrometry

  • LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
    • at 220 nm max. 0.002
    • at 254 nm max. 0.001
  • LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
    • Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ng/mL
  • Ultraviolet Absorbance (1.00-cm path vs water):
    • at 200 nm max. 0.05
    • at 220 nm max. 0.01
    • at 254 nm max. 0.01

Specifications

Melting Point -45.7°C
CAS 75-05-8
Density 0.782 g/cm³ (20°C)
Quantity 1 L
Flash Point 2°C
Packaging Glass Bottle
Residue after Evaporation max. 1 ppm
Assay Percent Range min. 99.9%
Molecular Formula C2H3N
MDL Number MFCD00001878
UN Number 1648
Water max. 0.01%
Synonym Methyl cyanide, Ethanoic acid nitrile, Ethanenitrile, Cyanomethane
Boiling Point 81.6°C (1013 hPa)
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.