missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, BAKER ANALYZED HPLC, J.T. Baker™

Catalog Number p-7138042
114.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
This item is not returnable. View return policy
75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N
This item is not returnable. View return policy
75-05-8
41.053
WEVYAHXRMPXWCK-UHFFFAOYSA-N
6342
CHEBI:38472
acetonitrile
CC#N

Get rapid, reproducible performance and separation results. Acetonitrile, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use
Melting Point -46°C
Absorbance Max. 0.5 at 200nm, Max. 0.3 at 210nm, Max. 0.2 at 220nm, Max. 0.05 at 254nm, Max. 0.01 at 280nm
Color Colorless
Molecular Formula C2H3N
Linear Formula CH3CN
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Formula Weight 41.05g/mol
Percent Purity ≥99.8%
Grade HPLC Isocratic Grade
Density 0.78g/cm3 at 20°C
Boiling Point 81°C
Quantity 1 L
Explosion Limit 4.4 - 16 Vol %
Flash Point 5°C
Ignition Point 524°C
Odor Aromatic
Packaging Glass
Physical Form Liquid
Refractive Index 1.344 n20/d
Residue after Evaporation Max. 0.0005%
Vapor Pressure 9.73 kPa (20°C)
Water Max. 0.1%
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'