missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile Anhydrous, Honeywell™ Riedel de-Haen
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Unit Size:
1 L
100mL
2.5 L
Description
- These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
- Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
- All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
- The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
- This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
Specifications
Specifications
| Density | 0.79 g/cm3 (20°C) |
| Quantity | 1 L |
| Flash Point | 12.8°C |
| Packaging | Septum-sealed Bottle |
| UN Number | 1648 |
| Formula Weight | 41.05 |
| Percent Purity | >99.9% |
| Chemical Name or Material | Acetonitrile |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction