missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, anhydrous, amine free, 99.9%

Product Code. 11395737
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395737 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11395737 Supplier Thermo Scientific Alfa Aesar Supplier No. 043440.K7
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75-05-8 | C2H3N | 41.053 g/mol

In peptide and DNA synthesis, HPLC.Acetonitrile is utilized as a polar aprotic solvent in organic synthesis and in the purification of butadiene. It is used as a mobile phase in HPLC and in LC-MS. Aqueous two-phase systems based on acetonitrile and carbohydrates play an important character in the extraction and purification of biomolecules called vanillins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -48°C to -45°C
CAS 75-05-8
Color Colorless
Density 0.786 g/cm3 at 20°C
Quantity 4 L
Flash Point 5°C (41°F)
Ignition Point 525°C (977°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.344
Assay Percent Range 99.9%
Molecular Formula C2H3N
Vapor Pressure 73mm Hg at 20°C
MDL Number MFCD00001878
Beilstein 741857
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Boiling Point 80°C to 82°C
Solubility Information Miscible with water and methanol.
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05
Chemical Name or Material Acetonitrile, Amine Free
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.