missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, For pesticide residue analysis, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
Specifications
Specifications
| Melting Point | -48°C (literature) |
| CAS Max % | 100.0000% |
| Density | 0.786 g/mL at 25 °C(lit.) |
| Boiling Point | 81.6°C |
| Quantity | 4 x 2.5 L |
| Flash Point | 6°C |
| Packaging | Glass Bottle |
| Linear Formula | CH3CN |
| UN Number | UN1648 |
| Beilstein | 741857 |
| Show More |
Safety and Handling
Product Identifier
- Acetonitrile
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : acetonitrile
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction