missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99.9%, Extra Dry, AcroSeal™, package of 4x25ML bottles

Product Code. 12317103 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
4 x 25 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12317103

Brand: Thermo Scientific Acros 448391000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 75-05-8 | C2H3N | 41.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -46.0°C
CAS 75-05-8
Color Colorless
Density 0.7810g/mL
Boiling Point 81.0°C to 82.0°C
Quantity 4 x 25 mL
Flash Point 2°C
Identification Pass Test
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Assay Percent Range 99.9%
Residue after Evaporation 0.0005 % max.
Specific Gravity 0.781
Molecular Formula C2H3N
Linear Formula CH3CN
MDL Number MFCD00001878
Viscosity 0.36 mPa.s (20°C)
Water 0.001% Max. (Coulometric)
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.053
PubChem CID 6342
ChEBI CHEBI:38472
Formula Weight 41.05
Percent Purity 99.9%
Grade Extra Dry
Chemical Name or Material Acetonitrile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.