missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile, 99.6%, ACS reagent

Product Code. 13167794 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
2.5 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
13167794 10 L 10 L
10444131 500 mL 500mL
10434131 1 L 1 L
10618414 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 13167794 Supplier Thermo Scientific Acros Supplier No. 423250100
  • ONLINE EXCLUSIVE PRICE
    £600.00
    £582.00 / 10 L
    Save £18.00
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 75-05-8 | C2H3N | 41.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.053
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym methyl cyanide, cyanomethane, ethanenitrile, methanecarbonitrile, ethyl nitrile, methane, cyano, acetonitrile, cyanure de methyl, methylkyanid, mecn
PubChem CID 6342
ChEBI CHEBI:38472
IUPAC Name acetonitrile
SMILES CC#N

Specifications

Melting Point -46.0°C
CAS Min % 99.5
CAS Max % 100.0
Decomposition Information >500°C
Density 0.7810g/mL
Quantity 10 L
Flash Point 2°C
Physical Form Liquid
Refractive Index 1.3430 to 1.3450
Residue after Evaporation 0.005% max.
Assay Percent Range 99.6%
Specific Gravity 0.781
Titratable Acid 0.008 meq/g max.
Titratable Base 0.0006 meq/g max.
Linear Formula CH3CN
Vapor Pressure Vapor Pressure: 97mbar at 20°C
Viscosity 0.36 mPa.s (20°C)
Water 0.3% max. (K.F.)
Beilstein 02, 183
Fieser 02,13; 12,450; 13,156; 15,1
Merck Index 15, 69
Boiling Point 81.0°C to 82.0°C
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol, ethyl acetate, acetone, ether
Formula Weight 41.05
Percent Purity 99.6%
Grade ACS
Chemical Name or Material Acetonitrile, For analysis ACS
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetonitrile
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.