missing translation for 'onlineSavingsMsg'
Learn More

Solstice Acetonitrile

Product Code. 15905480
Change view
Click to view available options
Quantity:
7 L
45 L
Unit Size:
45 L
7 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15905480 7 L 7 L
15995470 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15905480 Supplier Solstice Supplier No. 344427L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Acetonitrile for synthesis of DNA, ≥99.9% (GC).

  • For gas chromatography (GC).
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC.
  • Non-volatile matter and water content are specified for each solvent.

Chemical Identifiers

CAS 75-05-8
Molecular Formula C2H3N
Molecular Weight (g/mol) 41.05
MDL Number MFCD00001878
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
Synonym ACN, Methyl cyanide
IUPAC Name acetonitrile
SMILES CC#N

Specifications

pH Not applicable
Melting Point -46°C
Color Colorless
CAS Max % 100.00%
Density 0.7857 g/cm3 (20°C)
Quantity 7 L
Flash Point 6°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Linear Formula CH3CN
Vapor Pressure 97 hPa (20°C)
Viscosity No data available
UN Number UN1648
Beilstein 741857
Boiling Point 82°C (1.013 hPa)
Percent Purity ≥99.90%
Grade Biotech
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.