missing translation for 'onlineSavingsMsg'
Learn More

Solstice Acetonitrile

Product Code. 15905660
Change view
Click to view available options
Quantity:
200 L
Unit Size:
200 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15905660 200 L 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15905660 Supplier Solstice Supplier No. 33019200LRC
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acetonitrile, Puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Specifications

pH Not applicable
Melting Point -46°C
CAS 75-05-8
Color Colorless
CAS Max % 100.00%
Density 0.7857 g/cm3 (20°C)
Quantity 200 L
Flash Point 6°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Molecular Formula C2H3N
Linear Formula CH3CN
Vapor Pressure 97 hPa (20°C)
MDL Number MFCD00001878
Viscosity No data available
UN Number UN1648
Beilstein 741857
Concentration ≥99.50%
Synonym ACN, Methyl cyanide
Boiling Point 82°C (1.013 hPa)
InChI Key WEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILES CC#N
IUPAC Name acetonitrile
Molecular Weight (g/mol) 41.05
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Acetonitrile
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.