missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.1% formic acid, ≥99.5% (GC), Honeywell™ Riedel-de-Haën™ CHROMASOLV™

Product Code. 15634330
Click to view available options
Quantity:
1 L
4 x 4 L
Unit Size:
1 L
Pack of 4
This item is not returnable. View return policy

Product Code. 15634330

Brand: Honeywell Riedel-de Haën 346684X4L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

MDL Number MFCD00001878

Specifications

UN Number UN1648
Percent Purity ≥99.5% (GC)
Quantity 4 x 4 L
Flash Point 2°C (35.6°F)
Chemical Name or Material Acetonitrile with 0.1% formic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.