missing translation for 'onlineSavingsMsg'
Learn More

Acetonitrile with 0.05% TFA, LC/MS Grade, Thermo Scientific

Product Code. 30280964 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30280964 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30280964 Supplier Thermo Scientific Supplier No. TL01174000
 Restricted Item
Estimated Shipment: 28-04-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 76-05-1, 75-05-8
Quantity 4 L
Packaging Glass Bottle
Molecular Formula C2HF3O2
MDL Number MFCD00004169
InChI Key DTQVDTLACAAQTR-UHFFFAOYSA-N
SMILES OC(=O)C(F)(F)F
IUPAC Name trifluoroacetic acid
Molecular Weight (g/mol) 114.02
Grade LC-MS
Chemical Name or Material Acetonitrile with 0.05% TFA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.