missing translation for 'onlineSavingsMsg'
Learn More

Acetone, specified according to the requirements of Ph.Eur., Thermo Scientific Chemicals

Catalog Number p-7214371
19.37 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.


Click to view available options
Quantity:
1 L
5 L
25 L
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O
67-64-1
C3H6O
58.08
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
CC(C)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -95.0°C
Density 0.7900 g/mL
Boiling Point 56.0°C
Quantity 1 L
Flash Point -20°C
Packaging Glass Bottle
Refractive Index 1.3585
Specific Gravity 0.79
Linear Formula CH3COCH3
Viscosity 0.32 mPa.s (20°C)
Merck Index 15, 65
Solubility Information Solubility aq. soln.: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
Formula Weight 58.08
Chemical Name or Material Acetone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'