missing translation for 'onlineSavingsMsg'
Learn More

Acetone oxime, 98%

Product Code. 10346020 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346020 500 g Glass bottle 500g
10214560 100 g Glass bottle 100g
10606174 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10346020 Supplier Thermo Scientific Acros Supplier No. 102395000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 127-06-0 | C3H7NO | 73.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127-06-0
Molecular Formula C3H7NO
Molecular Weight (g/mol) 73.10
MDL Number MFCD00002118
InChI Key PXAJQJMDEXJWFB-UHFFFAOYSA-N
Synonym acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
PubChem CID 67180
ChEBI CHEBI:15349
IUPAC Name N-propan-2-ylidenehydroxylamine
SMILES CC(C)=NO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58.0°C to 62.0°C
Color White to Yellow
Boiling Point 135.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2C=NOH
Quantity 500 g
Beilstein 01, 649
Fieser 13,22
Merck Index 15, 73
Solubility Information Solubility in water: 330g/L (20°C). Other solubilities: soluble in alcohols and ethers
Formula Weight 73.09
Percent Purity 98%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material Acetone oxime, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Acetone oxime
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.