missing translation for 'onlineSavingsMsg'
Learn More

Acetone oxime, 98%, Thermo Scientific Chemicals

Catalog Number 11499722
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
127-06-0
C3H7NO
73.10
MFCD00002118
PXAJQJMDEXJWFB-UHFFFAOYSA-N
acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
67180
CHEBI:15349
N-propan-2-ylidenehydroxylamine
CC(C)=NO
This item is not returnable. View return policy
127-06-0
C3H7NO
73.10
MFCD00002118
PXAJQJMDEXJWFB-UHFFFAOYSA-N
acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5
67180
CHEBI:15349
N-propan-2-ylidenehydroxylamine
CC(C)=NO

It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an intermediate used in organic synthesis and in agriculture. It is also an important raw material.

Solubility
Soluble in water (330 g/L at 20°C).

Notes
Store away from oxidizing agents. Protect against electrostatic charges.
TRUSTED_SUSTAINABILITY
Melting Point 60°C to 63°C
Density 0.901
Boiling Point 133°C to 135°C
Flash Point 60°C (140°F)
Quantity 50 g
UN Number UN1325
Beilstein 1560146
Merck Index 14,75
Solubility Information Soluble in water (330g/L at 20°C).
Formula Weight 73.1
Percent Purity 98%
Chemical Name or Material Acetone oxime
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'