missing translation for 'onlineSavingsMsg'
Learn More

Acetone, MP Biomedicals™

Product Code. 11466221 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
0.5 L
1 L
4 L
Unit Size:
1 L
100mL
4 L
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466221 1 L 1 L
11442451 100 mL 100mL
11456221 0.5 L 500mL
11438262 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11466221 Supplier MP Biomedicals Supplier No. 0219383291
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acetone ACS, ≥99.5%, Clear, Colorless Liquid

  • It is a commonly used material in the manufacture of various chemicals like methyl isobutyl ketone, mesityl oxide, diacetone alcohol, acetic acid (via the ketene process), iodoform, and bromoform.
  • Other products where acetone is utilized in manufacture include rayon, photographic films, and paint and varnish removers.
  • In chemical synthesis, acetone has been utilized in the synthesis and isolation of both organic and inorganic compounds and complexes.
  • These include t-butanesulfinamide, terpenes, thiol-stabilized gold colloids, calixphyrin macrocycles, and trispyrazolylborate platinum compounds.
  • Acetone is also used in HPLC analysis of organic and inorganic species, including gold nanoparticle clusters and plant oils. Acetone has been utilized in protein crystallization.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

Melting Point -137°F
Color ≤10
Density 0.7905 to 0.793 g/cm3 (Lit.)
Quantity 1 L
Flash Point -17°C (1.4°F)
Physical Form Liquid
Refractive Index n20/D 1.359(lit.)
Titratable Base ≤0.0006 meq/g
Vapor Pressure 184 mmHg (20°C)
UN Number UN1090
Water ≤0.5%
Beilstein 635680EC
Synonym 2-propanone, dimethyl ketone, β-ketopropane
Boiling Point 56.2°C (at 1013 hPa (literature))
Solubility Information Soluble in water.
Grade ≥99.5%
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • Acetone, ACS
Palabra de advertencia
  • Danger
Clasificación
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Consejos de prudencia
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.