missing translation for 'onlineSavingsMsg'
Learn More

Acetone, Solstice

Product Code. 16605863
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Plastic Bottle
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
16605863 2.5 L Plastic Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605863 Supplier Fluka Supplier No. 40289H2.5L
  • ONLINE EXCLUSIVE PRICE
    £194.00
    £178.27 / 2.5 L
    Save £15.73
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Semiconductor grade VLSI ( 17617)

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories: Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications. Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Specifications

CAS 67-64-1
Quantity 2.5 L
Flash Point -17 °C
Packaging Plastic Bottle
Physical Form Liquid
Refractive Index 1.3580 - 1.3600
Molecular Formula C3H6O
MDL Number MFCD00008765
Water Max. 0.2 %
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
PubChem CID 180
ChEBI CHEBI:15347
Percent Purity min. 99.5 %
Grade Semiconductor
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.