missing translation for 'onlineSavingsMsg'
Learn More

Acetone-d6, 100% (Isotopic), Thermo Scientific™

Product Code. 11389927
Change view
Click to view available options
Quantity:
2 Ea.
Unit Size:
Pack of 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11389927 2 Ea. Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11389927 Supplier Thermo Scientific Alfa Aesar Supplier No. 042260.SA
Request Bulk or Custom Formats

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
MDL Number MFCD00044635
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C

Specifications

Melting Point -94°C
Density 0.872
Boiling Point 56°C
Flash Point -17°C (1°F)
Packaging Prescored ampoules, 0.75mL/ampoule
Refractive Index 1.3554
Quantity 2 Ea.
UN Number UN1090
Specific Gravity 0.872
Solubility Information Fully miscible in water.
Formula Weight 64.13
Percent Purity 100%
Physical Form Liquid
Chemical Name or Material Acetone-d6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.