missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1
£26.48 - £235.00
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Alert:
Repeated exposure may cause skin dryness or cracking.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15654510
|
Honeywell-Chromasolv
34850-1L |
1 L |
£26.48
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664510
|
Honeywell-Chromasolv
34850-2.5L |
2.5 L |
£57.99
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15684510
|
Honeywell-Chromasolv
34850-6X1L |
6 x 1 L |
£158.72
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15674510
|
Honeywell-Chromasolv
34850-4X2.5L |
4 x 2.5 L |
£235.00
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
-94°C | |
0.791g/mL at 25°C | |
n20/D 1.359 | |
CH3COCH3 | |
184mmHg (20°C) | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.8% | |
Acetone, for HPLC |
67-64-1 | |
56°C (760 mmHg) | |
C3H6O | |
MFCD00008765 | |
635680 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
CHROMASOLV™ |
Safety and Handling
P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
H225-H319-H336
missing translation for 'einecsNumber' : 200-662-2
missing translation for 'rtecsNumber' : AL3150000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Acetone
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.