missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
147.51 GBP valid until 2024-12-31
Use promo code "21898" to get your promotional price.
147.51 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.
Use promo code "21898" to get your promotional price.
147.51 GBP valid until 2024-12-31
Use promo code "21905" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Specifications
Melting Point | -94°C |
Density | 0.791g/mL at 25°C |
Boiling Point | 56°C (760 mmHg) |
Quantity | 4 x 2.5 L |
Refractive Index | n20/D 1.359 |
Linear Formula | CH3COCH3 |
Vapor Pressure | 184mmHg (20°C) |
Beilstein | 635680 |
Formula Weight | 58.08g/mol |
Percent Purity | ≥99.8% |
Show More |
Safety and Handling
Product Identifier
- Acetone
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- EUH066-Repeated exposure may cause skin dryness or cracking.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'