missing translation for 'onlineSavingsMsg'
Learn More

Acetone, BAKER ANALYZED™, HPLC, J.T. Baker™

Product Code. 10281291 Shop All J.T.Baker Products
Change view
Click to view available options
:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10281291 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10281291 Supplier JT Baker Supplier No. 8142.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For HPLC

  • Ultraviolet Absorbance (1.00-cm path vs water):
    • at 330 nm max. 1.00
    • at 340 nm max. 0.10
    • at 350 nm max. 0.01
    • at 400 nm max. 0.01
    • UV Cut-off, nm max. 330

Specifications

Melting Point -95.4°C
CAS 67-64-1
Color Max. 10 (APHA)
Density 0.792 g/cm³ (20°C)
Quantity 2.5 L
Residue after Evaporation max. 2 ppm
Assay Percent Range min. 99.7%
Titratable Acid max. 0.0003 meq/g
Molecular Formula C3H6O
Titratable Base max. 0.0006 meq/g
MDL Number MFCD00008765
Water max. 0.2%
Synonym Propanone
Boiling Point 56.2°C (1013 hPa)
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.