missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS/USP, Greenfield

Product Code. 30286168
Change view
Click to view available options
Packaging:
4x1 Fluorinated Poly Gallon Case
55 Gallon Metal Drum
55 Gallon Poly Drum
Unit Size:
15.14 L
208.2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
30286168 4x1 Fluorinated Poly Gallon Case 15.14 L
30286189 55 Gallon Poly Drum 208.2 L
30286188 55 Gallon Metal Drum 208.2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30286168 Supplier Pharmco Supplier No. 329000000CSGF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1

Specifications

Quantity 4x1 Fluorinated Poly Gallon Case
Packaging 4x1 Fluorinated Poly Gallon Case
Boiling Point 56°C
Grade Reagent
Chemical Name or Material Acetone, ACS/USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.