missing translation for 'onlineSavingsMsg'
Learn More

Acetone, ACS/USP-Grade, 5, Pharmco

Product Code. 30286183
Change view
Click to view available options
Unit Size:
18.93 L
Packaging:
5 Gallon Poly Pail
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Packaging
30286183 18.93 L -
30286177 18.93 L 5 Gallon Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30286183 Supplier Pharmco Supplier No. 329000000PL05M
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-64-1

Specifications

Quantity 5 Gallon
Molecular Formula C3H6O
MDL Number MFCD00008765
Boiling Point 56°C
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
SMILES CC(C)=O
IUPAC Name propan-2-one
Molecular Weight (g/mol) 58.08
Grade Reagent
Chemical Name or Material Acetone, ACS/USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.