missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetone, Histological Grade, ≥99.5%, Honeywell
£24.93 - £237.71
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Description
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
-94°C (literature) | |
≥90.0000% | |
0.791 g/mL at 25 °C(lit.) | |
0.132 | |
1.359 | |
CH3COCH3 | |
184mmHg (20°C) | |
635680 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
Histological |
67-64-1 | |
≤100.0000% | |
55°C to 57°C | |
−17°C | |
C3H6O | |
MFCD00008765 | |
UN1090 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
>99.5% | |
Acetone, Histological Grade, ≥99.5% |
Safety and Handling
P210-P305 + P351 + P338-P370 + P378-P403 + P235
missing translation for 'shelfLife' : 1080 days from date of manufacture
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Acetone
missing translation for 'einecsNumber' : 200-662-2
missing translation for 'rtecsNumber' : 200-662-2
Recommended Storage : Room Temp