missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Acetone

Product Code. 15945650
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15945650 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15945650 Supplier Riedel-de Haën Supplier No. 2420118L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Acetone, Puriss., meets analytical specification of BP, NF, Ph. Eur., ≥99% (GC).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Chemical Identifiers

CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym 2-Propanone, Dimethyl Ketone
IUPAC Name propan-2-one
SMILES CC(C)=O

Specifications

pH Not applicable
Melting Point -95°C
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.79 g/cm3 (20°C)
Quantity 18 L
Flash Point -17°C
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss.
Linear Formula CH3COCH3
Vapor Pressure 240 hPa (20°C)
Viscosity 0.32 mPa.s (20°C)
UN Number UN1090
Beilstein 635680
Boiling Point 55°C to 57°C (1.013 hPa)
Percent Purity ≥99.00%
Grade Pharmacopeia
Chemical Name or Material Acetone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.