missing translation for 'onlineSavingsMsg'
Learn More

Acetoin, 96%, dimer

Product Code. 10196840 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10196840

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23147-57-1 | C8H16O4 | 176.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23147-57-1
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
MDL Number MFCD00038696
InChI Key DFMGATPNJMFDCR-UHFFFAOYNA-N
Synonym acetoin, 3-hydroxy-2-butanone, acetyl methyl carbinol, dimethylketol, 2-butanone, 3-hydroxy, acetylmethylcarbinol, 2,3-butanolone, 2-hydroxy-3-butanone, 1-hydroxyethyl methyl ketone, methanol, acetylmethyl
PubChem CID 179
ChEBI CHEBI:15688
IUPAC Name 3-hydroxybutan-2-one
SMILES CC1OC(C)(O)C(C)OC1(C)O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 15.0°C
Color Green-Yellow to Yellow or White to Yellow
Density 1.0000g/mL
Boiling Point 148.0°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH(OH)CH3
Refractive Index 1.4171
Quantity 100 g
Beilstein 01, 827
Merck Index 15, 63
Specific Gravity 1
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,propylene glycol,insoluble in vegetable oil
Formula Weight 88.11
Percent Purity 96%
Physical Form Liquid (Monomer) or Powder or Crystals (Dimer)
Chemical Name or Material Acetoin, pract., 93%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetoin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.