missing translation for 'onlineSavingsMsg'
Learn More

Acetoin (~90%), TRC

Product Code. 30011040
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
Unit Size:
25g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011040 5 g 5g
30005391 50 g 50g
30006026 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011040 Supplier Toronto Research Chemicals Supplier No. TRCA1637785G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-86-0
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
Synonym 2-Butanone, 3-hydroxy-, 3-Hydroxy-2-butanone, (±)-3-Hydroxybutan-2-one, (±)-Acetoin, 1-Hydroxyethyl methyl ketone, 2,3-Butanolone, 2-Hydroxy-3-butanone, 3-Hydroxybutanone, 3-Hydroxyl-2-butanone, 3-Oxo-2-butanol, Acetoin, Acetoine, Acetyl methyl carbinol, DL-Acetoin, Dimethylketol, Methanol, acetylmethyl-, NSC 7609, γ-Hydroxy-β-oxobutane
IUPAC Name 3-hydroxybutan-2-one
SMILES CC(O)C(=O)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C4H8O2/c1-3(5)4(2)6/h3,5H,1-2H3
Formula Weight 88.0524
Chemical Name or Material Acetoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.