missing translation for 'onlineSavingsMsg'
Learn More

Acetohydroxamic acid, MedChemExpress

Product Code. 16488696
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488696 100 mg 100mg
16412266 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16488696 Supplier Medchem Express Supplier No. HYB1235
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acetohydroxamic acid is a potent and irreversible inhibitor of bacterial and plant urease and also used as adjunctive therapy in chronic urinary infection.

Acetohydroxamic acid is a potent and irreversible inhibitor of bacterial and plant urease and also used as adjunctive therapy in chronic urinary infection.

Specifications

Chemical Name or Material Acetohydroxamic acid
Physical Form Powder
CAS 546-88-3
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C2H5NO2
For Use With (Application) COVID-19-immunoregulation
Synonym AHA
Solubility Information DMSO : 250 mg/mL (3330.23 mM; Need ultrasonic) ∣H2O : 100 mg/mL (1332.09 mM; Need ultrasonic)
SMILES CC(NO)=O
Molecular Weight (g/mol) 75.07
Formula Weight 75.07
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.