missing translation for 'onlineSavingsMsg'
Learn More

Acetohydrazide, TRC

Product Code. 30133645
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133645 5 g 5g
30041138 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133645 Supplier Toronto Research Chemicals Supplier No. TRCA1637705G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-57-1
Molecular Formula C2H6N2O
Molecular Weight (g/mol) 74.08
Synonym Acetic Acid Hydrazide, Acethydrazide, Acetic hydrazide, Acetohydrazine, Acetyl hydrazide, Acetylhydrazine, ENT-61241, Ethanehydrazonic Acid, Monoacetylhydrazine, NSC 2271, NSC 53155
IUPAC Name acetohydrazide
SMILES CC(=O)NN

Specifications

Quantity 5 g
InChI Formula InChI=1S/C2H6N2O/c1-2(5)4-3/h3H2,1H3,(H,4,5)
Formula Weight 74.048
Chemical Name or Material Acetohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.