missing translation for 'onlineSavingsMsg'
Learn More

Acetohexamide, MedChemExpress

Product Code. 16492216
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492216 500 mg 500mg
16468476 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16492216 Supplier Medchem Express Supplier No. HYB0881
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Acetohexamide is a first-generation sulfonylurea medication used to treat diabetes mellitus type 2; stimulate the pancreas to secrete insulin.

Acetohexamide is a first-generation sulfonylurea medication used to treat diabetes mellitus type 2

Specifications

Chemical Name or Material Acetohexamide
CAS 968-81-0
Color White
Purity Grade Notes Research
Molecular Formula C15H20N2O4S
Quantity 500 mg
Solubility Information DMSO : 25 mg/mL (77.07 mM; Need ultrasonic)
SMILES O=S(C1=CC=C(C(C)=O)C=C1)(NC(NC2CCCCC2)=O)=O
Molecular Weight (g/mol) 324.4
Formula Weight 324.4
Percent Purity 99.39%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.