missing translation for 'onlineSavingsMsg'
Learn More

Acetobromo-alpha-D-glucuronic Acid Methyl Ester (contains 2% CaCO3), TRC

Product Code. 30003983
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003983 25 g 25g
30009448 10 g 10g
17941065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003983 Supplier Toronto Research Chemicals Supplier No. TRCA16200025G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21085-72-3
Molecular Formula C13 H17 Br O9
Molecular Weight (g/mol) 397.17
IUPAC Name methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate
SMILES COC(=O)[C@H]1O[C@H](Br)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C13H17BrO9/c1-5(15)20-8-9(21-6(2)16)11(13(18)19-4)23-12(14)10(8)22-7(3)17/h8-12H,1-4H3/t8-,9-,10+,11-,12-/m0/s1
Formula Weight 396.0056
Chemical Name or Material Acetobromo-a-D-glucuronic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.